Gene Transcripts

Gene Symbol
Smyd3
Entrez Gene ID
498295
Gene Name
SET and MYND domain containing 3
Gene Aliases
Chromosome Location
Chr. 13: 97330120 - 97566717 on Build Rnor_6.0
UniGene ID
Rn.28284
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025762.1 NP_001020933.1 558
GenBank mRNA BC097455.1 - 558

View Full Product Details