Gene Transcripts

Gene Symbol
Zfp839
Entrez Gene ID
500723
Gene Name
zinc finger protein 839
Gene Aliases
RGD1562557, Znf839
Chromosome Location
Chr. 6: 135269855 - 135287371 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006240613.2 XP_006240675.1 8 3642
XM_006240614.2 XP_006240676.1 7 3414
XM_006240615.2 XP_006240677.1 8 3137
XM_006240616.2 XP_006240678.2 8 3846
XM_008764973.1 XP_008763195.1 7 3619

View Full Product Details