Gene Transcripts

Gene Symbol
Lrch2
Entrez Gene ID
680591
Gene Name
leucine-rich repeats and calponin homology (CH) domain containing 2
Gene Aliases
Chromosome Location
Chr. X: 118532396 - 118615798 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001057911.4 XP_001057911.2 3 590
XM_006257439.2 XP_006257501.1 3 590

View Full Product Details