Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142418.1 | NP_001135890.1 | 5 | 1188 | |
NM_001142419.1 | NP_001135891.1 | 4 | 1121 | ||
NM_001142420.1 | NP_001135892.1 | 3 | 968 | ||
NM_001142421.1 | NP_001135893.1 | 4 | 1070 | ||
NM_001142422.1 | NP_001135894.1 | 5 | 1170 | ||
NM_001142423.1 | NP_001135895.1 | 4 | 1088 | ||
NM_001142424.1 | NP_001135896.1 | 5 | 1137 | ||
NM_001142425.1 | NP_001135897.1 | 4 | 1108 | ||
NM_001142426.1 | NP_001135898.1 | 3 | 986 | ||
NM_001142427.1 | NP_001135899.1 | 4 | 1090 | ||
NM_001142428.1 | NP_001135900.1 | 5 | 1119 | ||
NM_001142429.1 | NP_001135901.1 | 5 | 1157 | ||
NM_001142430.1 | NP_001135902.1 | 4 | 1035 | ||
NM_001142431.1 | NP_001135903.1 | 4 | 1017 | ||
NM_001142432.1 | NP_001135904.1 | 5 | 1139 | ||
NM_012286.2 | NP_036418.1 | 4 | 1139 | ||
GenBank mRNA | AF100620.1 | - | 4 | 989 | |
AF167174.1 | - | 4 | 984 | ||
AK056012.1 | - | 4 | 976 | ||
AK095298.1 | - | 2 | 1901 | ||
AK126115.1 | - | 6 | 964 | ||
AK126427.1 | - | 4 | 1916 | ||
AK223504.1 | - | 4 | 1020 | ||
AK302083.1 | - | 5 | 920 | ||
BC056899.1 | - | 5 | 974 | 3905425 | |
BC093013.1 | - | 5 | 1018 | 30528536 | |
D14812.1 | - | 4 | 989 |