Gene Transcripts

Gene Symbol
Mrps16
Entrez Gene ID
688912
Gene Name
mitochondrial ribosomal protein S16
Gene Aliases
Chromosome Location
Chr. 15: 4351292 - 4353694 on Build Rnor_6.0
UniGene ID
Rn.8846
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109518.1 NP_001102988.1 2 250
GenBank mRNA FQ210083.1 - 2 205
FQ228795.1 - 2 228

View Full Product Details