Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 16 | 1441 | |
XM_011534325.2 | XP_011532627.1 | 16 | 1422 | ||
XM_011534327.1 | XP_011532629.1 | 16 | 1422 | ||
XM_011534328.2 | XP_011532630.1 | 16 | 1422 | ||
XM_011534329.1 | XP_011532631.1 | 16 | 1422 | ||
XM_011534330.2 | XP_011532632.1 | 16 | 1422 | ||
XM_017007595.1 | XP_016863084.1 | 14 | 1190 | ||
XM_017007596.1 | XP_016863085.1 | 16 | 1467 | ||
XM_017007597.1 | XP_016863086.1 | 8 | 756 | ||
XM_017007598.1 | XP_016863087.1 | 16 | 1422 | ||
XM_017007599.1 | XP_016863088.1 | 15 | 1325 | ||
XM_017007600.1 | XP_016863089.1 | 13 | 1235 | ||
GenBank mRNA | AB015046.1 | - | 16 | 1461 | |
AK001205.1 | - | 9 | 712 | ||
AK025728.1 | - | 16 | 1393 | ||
AK293325.1 | - | 14 | 1195 | ||
AK314386.1 | - | 16 | 1389 | ||
BC137076.1 | - | 16 | 1373 | 9021073 | |
BC137080.1 | - | 16 | 1373 | 9021077 |