Gene Transcripts

Gene Symbol
Lpar6
Entrez Gene ID
691774
Gene Name
lysophosphatidic acid receptor 6
Gene Aliases
P2ry5
Chromosome Location
Chr. 15: 55126953 - 55128762 on Build Rnor_6.0
UniGene ID
Rn.203876
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045843.1 NP_001039308.1 1 1503
GenBank mRNA BC098703.1 - 1 1503

View Full Product Details