Gene Transcripts

Gene Symbol
NEU2
Entrez Gene ID
4759
Gene Name
neuraminidase 2
Gene Aliases
SIAL2
Chromosome Location
Chr. 2: 233032672 - 233035057 on Build GRCh38
UniGene ID
Hs.532681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005383.2 NP_005374.2 2 1076
GenBank mRNA BC069151.1 - 2 1076
BC107053.2 - 2 1076 40004620

View Full Product Details