Gene Transcripts
- Gene Symbol
- DUX5
- Entrez Gene ID
- 26581
- Gene Name
- double homeobox 5
- Gene Aliases
- DUX1
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012149.2 |
NP_036281.2
|
1 |
250 |
|
- Gene Symbol
- DUX1
- Entrez Gene ID
- 26584
- Gene Name
- double homeobox 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.274469
- Species
- Homo sapiens
- Gene Symbol
- DUX3
- Entrez Gene ID
- 26582
- Gene Name
- double homeobox 3
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012148.2 |
NP_036280.2
|
1 |
250 |
|
View Full Product Details