Gene Transcripts

Gene Symbol
LCE3A
Entrez Gene ID
353142
Gene Name
late cornified envelope 3A
Gene Aliases
LEP13
Chromosome Location
Chr. 1: 152622834 - 152623103 on Build GRCh38
UniGene ID
Hs.553714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178431.1 NP_848518.1 1 149
GenBank mRNA BC120933.1 - 1 149
BC120934.1 - 1 149

View Full Product Details