Gene Transcripts
- Gene Symbol
- CT47A4
- Entrez Gene ID
- 728075
- Gene Name
- cancer/testis antigen family 47, member A4
- Gene Aliases
- CT47.4
- Chromosome Location
- Chr. X: 120967886 - 120971306 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A1
- Entrez Gene ID
- 728096
- Gene Name
- cancer/testis antigen family 47, member A1
- Gene Aliases
- CT47.1
- Chromosome Location
- Chr. X: 120982466 - 120985886 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A10
- Entrez Gene ID
- 728036
- Gene Name
- cancer/testis antigen family 47, member A10
- Gene Aliases
- CT47, CT47.10
- Chromosome Location
- Chr. X: 120938701 - 120942121 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A11
- Entrez Gene ID
- 255313
- Gene Name
- cancer/testis antigen family 47, member A11
- Gene Aliases
- CT47.11
- Chromosome Location
- Chr. X: 120933840 - 120937260 on Build GRCh38
- UniGene ID
- Hs.522775
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173571.2 |
NP_775842.2
|
1, 2 |
1008 |
|
GenBank mRNA |
BC029540.1 |
- |
1, 2 |
1012 |
|
- Gene Symbol
- CT47A12
- Entrez Gene ID
- 100507170
- Gene Name
- cancer/testis antigen family 47, member A12
- Gene Aliases
- CT47, CT47.12
- Chromosome Location
- Chr. X: 120877490 - 120932399 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A2
- Entrez Gene ID
- 728090
- Gene Name
- cancer/testis antigen family 47, member A2
- Gene Aliases
- CT47.2
- Chromosome Location
- Chr. X: 120977606 - 120981026 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A3
- Entrez Gene ID
- 728082
- Gene Name
- cancer/testis antigen family 47, member A3
- Gene Aliases
- CT47.3
- Chromosome Location
- Chr. X: 120972746 - 120976166 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A5
- Entrez Gene ID
- 728072
- Gene Name
- cancer/testis antigen family 47, member A5
- Gene Aliases
- CT47.5
- Chromosome Location
- Chr. X: 120963026 - 120966446 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A6
- Entrez Gene ID
- 728062
- Gene Name
- cancer/testis antigen family 47, member A6
- Gene Aliases
- CT47.6
- Chromosome Location
- Chr. X: 120958165 - 120961588 on Build GRCh38
- UniGene ID
- Hs.723061
- Species
- Homo sapiens
- Gene Symbol
- CT47A7
- Entrez Gene ID
- 653282
- Gene Name
- cancer/testis antigen family 47, member A7
- Gene Aliases
- CT47.7
- Chromosome Location
- Chr. X: 120953282 - 120956600 on Build GRCh38
- UniGene ID
- Hs.712504
- Species
- Homo sapiens
- Gene Symbol
- CT47A8
- Entrez Gene ID
- 728049
- Gene Name
- cancer/testis antigen family 47, member A8
- Gene Aliases
- CT47.8
- Chromosome Location
- Chr. X: 120948422 - 120951842 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47A9
- Entrez Gene ID
- 728042
- Gene Name
- cancer/testis antigen family 47, member A9
- Gene Aliases
- CT47.9
- Chromosome Location
- Chr. X: 120943561 - 120946981 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
- Gene Symbol
- CT47B1
- Entrez Gene ID
- 643311
- Gene Name
- cancer/testis antigen family 47, member B1
- Gene Aliases
- CT47.13, CT47A13
- Chromosome Location
- Chr. X: 120872554 - 120875925 on Build GRCh38
- UniGene ID
- Hs.729566
- Species
- Homo sapiens
View Full Product Details