Gene Transcripts

Gene Symbol
ANKRD34C-AS1
Entrez Gene ID
729911
Gene Name
ANKRD34C antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 15: 79191707 - 79283945 on Build GRCh38
UniGene ID
Hs.513931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038997.1 2 337
GenBank mRNA DA316055.1 - 2 338
DA323807.1 - 2 301
DA336675.1 - 2 217
DA364073.1 - 2 218

View Full Product Details