Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081085.2 | NP_001074554.1 | 7 | 2340 | |
NM_001290407.1 | NP_001277336.1 | 6 | 2238 | ||
GenBank mRNA | AB197927.1 | - | 6 | 2238 | |
AK154143.1 | - | 7 | 2052 | ||
AK163973.1 | - | 7 | 1559 | ||
AK164615.1 | - | 8 | 1800 | ||
AK166625.1 | - | 7 | 2015 |