Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007006.2 | NP_008937.1 | 6 | 741 | |
GenBank mRNA | AJ001810.1 | - | 6 | 594 | |
AK312721.1 | - | 6 | 744 | ||
AL535307.3 | - | 6 | 741 | ||
BC001403.2 | - | 6 | 702 | 3139379 | |
BX537360.1 | - | 6 | 745 | ||
CR456919.1 | - | 6 | 594 |