Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006248358.2 | XP_006248420.1 | 3 | 247 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001105881.1 | NP_001099351.1 | 488 | ||
XM_006248326.2 | XP_006248388.1 | 3 | 494 | ||
GenBank mRNA | BC169000.1 | - | 3 | 484 | |
FQ216855.1 | - | 3 | 404 |