Gene Transcripts

Gene Symbol
Gpr87
Entrez Gene ID
310443
Gene Name
G protein-coupled receptor 87
Gene Aliases
Chromosome Location
Chr. 2: 149398949 - 149405195 on Build Rnor_6.0
UniGene ID
Rn.218426
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107677.1 NP_001101147.1 1 223
XM_006232401.2 XP_006232463.1 2 856

View Full Product Details