Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000301.3 | NP_000292.1 | 9 | 1187 | |
GenBank mRNA | AK298338.1 | - | 5 | 543 | |
AK308036.1 | - | 7 | 732 | ||
AK312966.1 | - | 9 | 1138 | ||
BC060513.1 | - | 9 | 1113 | 4609795 | |
K02922.1 | - | 2 | 202 | ||
M74220.1 | - | 9 | 1124 | ||
X05199.1 | - | 9 | 1129 |