Gene Transcripts

Gene Symbol
Olr836
Entrez Gene ID
298394
Gene Name
olfactory receptor 836
Gene Aliases
Chromosome Location
Chr. 5: 59265334 - 59266293 on Build Rnor_6.0
UniGene ID
Rn.143213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000406.1 NP_001000406.1 1 601

View Full Product Details