Gene Transcripts
- Gene Symbol
- SIRT5
- Entrez Gene ID
- 23408
- Gene Name
- sirtuin 5
- Gene Aliases
- SIR2L5
- Chromosome Location
- Chr. 6: 13574222 - 13615158 on Build GRCh38
- UniGene ID
- Hs.567431
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193267.2 |
NP_001180196.1
|
5, 6 |
763 |
|
NM_001242827.1 |
NP_001229756.1
|
4, 5 |
629 |
|
NM_012241.4 |
NP_036373.1
|
5, 6 |
785 |
|
NM_031244.3 |
NP_112534.1
|
5, 6 |
763 |
|
XM_005248968.4 |
XP_005249025.1
|
5, 6 |
700 |
|
XM_011514444.2 |
XP_011512746.1
|
6, 7 |
834 |
|
XM_017010620.1 |
XP_016866109.1
|
4, 5 |
663 |
|
XM_017010621.1 |
XP_016866110.1
|
5, 6 |
700 |
|
XM_017010622.1 |
XP_016866111.1
|
6, 7 |
893 |
|
XM_017010623.1 |
XP_016866112.1
|
6, 7 |
1023 |
|
XM_017010624.1 |
XP_016866113.1
|
4, 5 |
663 |
|
XM_017010625.1 |
XP_016866114.1
|
4, 5 |
663 |
|
GenBank mRNA |
AF083110.1 |
- |
5, 6 |
746 |
|
AJ420496.1 |
- |
5, 6 |
692 |
|
AK000355.1 |
- |
5, 6 |
691 |
|
AK294162.1 |
- |
4, 5 |
557 |
|
AK302467.1 |
- |
5, 6 |
698 |
|
BC000126.1 |
- |
5, 6 |
769 |
3352592 |
BX538030.1 |
- |
4, 5 |
576 |
|
View Full Product Details