Gene Transcripts

Gene Symbol
NNT
Entrez Gene ID
23530
Gene Name
nicotinamide nucleotide transhydrogenase
Gene Aliases
GCCD4
Chromosome Location
Chr. 5: 43602574 - 43705566 on Build GRCh38
UniGene ID
Hs.482043
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012343.3 NP_036475.3 18 2999
NM_182977.2 NP_892022.2 18 2965
XM_005248274.4 XP_005248331.1 18 2977
XM_005248275.4 XP_005248332.1 13 2227
XM_006714461.3 XP_006714524.1 17 2591
XM_011514001.2 XP_011512303.1 18 2996
XM_011514002.2 XP_011512304.1 17 2686
XM_017009293.1 XP_016864782.1 18 2867
GenBank mRNA AK297558.1 - 11 1612
AK299622.1 - 10 1429
AL831822.2 - 18 2966
BC032370.1 - 2 194
BC110543.1 - 18 2815 40036686
BC110544.1 - 17 2812 40036689
U40490.1 - 18 2887
Z50101.1 - 17 2787

View Full Product Details