Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 2 | 356 | |
NM_013241.2 | NP_037373.2 | 2 | 356 | ||
XM_011523043.2 | XP_011521345.1 | 2 | 572 | ||
XM_011523044.1 | XP_011521346.1 | 2 | 301 | ||
XM_011523045.2 | XP_011521347.1 | 2 | 66 | ||
GenBank mRNA | AB041046.1 | - | 2 | 326 | |
AB209584.1 | - | 2 | 298 | ||
AF113615.1 | - | 2 | 262 | ||
AK304232.1 | - | 2 | 312 | ||
AY192154.1 | - | 2 | 244 | ||
BC033084.1 | - | 2 | 301 | 5456031 | |
CN302043.1 | - | 2 | 358 | ||
KJ658417.1 | - | 2 | 244 |