Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001311182.1 | NP_001298111.1 | 6 | 772 | |
NM_022046.5 | NP_071329.2 | 6 | 772 | ||
XM_017026820.1 | XP_016882309.1 | 5 | 1130 | ||
GenBank mRNA | AF283670.1 | - | 5 | 631 | |
BC074904.2 | - | 5 | 588 | 30915253 | |
BC074905.2 | - | 5 | 588 | 30915482 | |
BC114614.1 | - | 6 | 772 | 40031928 |