Gene Transcripts
- Gene Symbol
- SLC25A37
- Entrez Gene ID
- 51312
- Gene Name
- solute carrier family 25 member 37
- Gene Aliases
- HT015, MFRN, MFRN1, MSC, MSCP, PRO1278, PRO1584, PRO2217
- Chromosome Location
- Chr. 8: 23528795 - 23575463 on Build GRCh38
- UniGene ID
- Hs.596025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317812.1 |
NP_001304741.1
|
1, 2 |
411 |
|
NM_016612.3 |
NP_057696.2
|
1, 2 |
411 |
|
GenBank mRNA |
AF223466.1 |
- |
1, 2 |
247 |
|
AK074708.1 |
- |
1, 2 |
250 |
|
AK075313.1 |
- |
1, 2 |
176 |
|
AK223194.1 |
- |
1, 2 |
252 |
|
AL833186.1 |
- |
1, 2 |
153 |
|
AY032628.1 |
- |
1, 2 |
247 |
|
BC015013.2 |
- |
1, 2 |
216 |
|
BC132799.1 |
- |
1, 2 |
275 |
40146821 |
BC132801.1 |
- |
1, 2 |
275 |
40146823 |
BI460337.1 |
- |
1, 2 |
365 |
|
BM926580.1 |
- |
1, 2 |
254 |
|
DB057758.1 |
- |
1, 2 |
412 |
|
View Full Product Details