Gene Transcripts
- Gene Symbol
- NUDT9
- Entrez Gene ID
- 53343
- Gene Name
- nudix hydrolase 9
- Gene Aliases
- NUDT10
- Chromosome Location
- Chr. 4: 87420002 - 87459454 on Build GRCh38
- UniGene ID
- Hs.149500
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001248011.1 |
NP_001234940.1
|
5, 6 |
1020 |
|
NM_024047.4 |
NP_076952.1
|
6, 7 |
1116 |
|
NM_198038.2 |
NP_932155.1
|
6, 7 |
758 |
|
XM_011532032.2 |
XP_011530334.1
|
6, 7 |
3295 |
|
GenBank mRNA |
AF265341.1 |
- |
6, 7 |
757 |
|
AF273028.1 |
- |
6, 7 |
1080 |
|
AK027295.1 |
- |
6, 7 |
1106 |
|
AK074845.1 |
- |
6, 7 |
758 |
|
AK075408.1 |
- |
6, 7 |
1036 |
|
AK308028.1 |
- |
5, 6 |
787 |
|
AY026252.1 |
- |
6, 7 |
1111 |
|
AY359123.1 |
- |
6, 7 |
810 |
|
BC000542.2 |
- |
6, 7 |
876 |
3163944 |
BI552154.1 |
- |
|
761 |
|
View Full Product Details