Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142928.1 | NP_001136400.1 | 3 | 1570 | |
NM_023942.2 | NP_076431.1 | 2 | 1400 | ||
XM_006716095.2 | XP_006716158.1 | 3 | 1790 | ||
GenBank mRNA | AF161413.1 | - | 1 | 291 | |
AK025869.1 | - | 2 | 1072 | ||
AK055954.1 | - | 3 | 1486 | ||
AK093915.1 | - | 1 | 2120 | ||
AK098061.1 | - | 3 | 1576 | ||
BC001354.1 | - | 2 | 1099 | 3163787 |