Gene Transcripts
- Gene Symbol
- ZNF768
- Entrez Gene ID
- 79724
- Gene Name
- zinc finger protein 768
- Gene Aliases
- Chromosome Location
- Chr. 16: 30524001 - 30532367 on Build GRCh38
- UniGene ID
- Hs.85658
- Species
- Homo sapiens
- Gene Symbol
- ZNF747
- Entrez Gene ID
- 65988
- Gene Name
- zinc finger protein 747
- Gene Aliases
- Chromosome Location
- Chr. 16: 30530367 - 30534970 on Build GRCh38
- UniGene ID
- Hs.592032
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK299308.1 |
- |
4 |
1458 |
|
View Full Product Details