Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001165928.3 | NP_001159400.2 | 6 | 2636 | |
NM_001177634.2 | NP_001171105.1 | 6 | 2579 | ||
NM_001177635.2 | NP_001171106.1 | 4 | 2168 | ||
NM_001177636.2 | NP_001171107.1 | 5 | 2557 | ||
NM_001177637.2 | NP_001171108.1 | 4 | 2395 | ||
NM_001177638.2 | NP_001171109.1 | 4 | 2426 | ||
NM_001177639.2 | NP_001171110.1 | 3 | 2116 | ||
NM_001177640.2 | NP_001171111.1 | 4 | 2332 | ||
NM_001177641.2 | NP_001171112.1 | 4 | 2422 | ||
NM_001177642.2 | NP_001171113.1 | 4 | 2327 | ||
NM_001177643.2 | NP_001171114.1 | 3 | 2326 | ||
NM_001177644.2 | NP_001171115.1 | 3 | 2415 | ||
NM_004393.5 | NP_004384.4 | 3 | 2275 | ||
GenBank mRNA | AK291692.1 | - | 3 | 2117 | |
BC012740.2 | - | 3 | 2210 | 4099382 | |
BC014616.2 | - | 3 | 2210 | 2964740 | |
L19711.1 | - | 3 | 2251 |