Gene Transcripts
- Gene Symbol
- C15orf41
- Entrez Gene ID
- 84529
- Gene Name
- chromosome 15 open reading frame 41
- Gene Aliases
- HH114
- Chromosome Location
- Chr. 15: 36579603 - 36810260 on Build GRCh38
- UniGene ID
- Hs.48348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130010.2 |
NP_001123482.1
|
9, 10 |
876 |
|
NM_001290232.1 |
NP_001277161.1
|
9, 10 |
567 |
|
NM_001290233.1 |
NP_001277162.1
|
9, 10 |
876 |
|
NM_001321756.1 |
NP_001308685.1
|
8, 9 |
830 |
|
NM_001321757.1 |
NP_001308686.1
|
10, 11 |
910 |
|
NM_001321758.1 |
NP_001308687.1
|
7, 8 |
765 |
|
NM_001321759.1 |
NP_001308688.1
|
9, 10 |
876 |
|
NM_032499.5 |
NP_115888.1
|
10, 11 |
622 |
|
XM_011522111.1 |
XP_011520413.1
|
9, 10 |
868 |
|
XM_017022676.1 |
XP_016878165.1
|
6, 7 |
670 |
|
XM_017022677.1 |
XP_016878166.1
|
6, 7 |
670 |
|
GenBank mRNA |
AF114263.1 |
- |
10, 11 |
597 |
|
AK026504.1 |
- |
9, 10 |
845 |
|
AK315446.1 |
- |
9, 10 |
536 |
|
BC006254.2 |
- |
9, 10 |
635 |
3952242 |
BC042750.1 |
- |
1, 2 |
4981 |
|
BX647650.1 |
- |
3, 4 |
246 |
|
DA534917.1 |
- |
8, 9 |
531 |
|
View Full Product Details