Gene Transcripts

Gene Symbol
ZNF383
Entrez Gene ID
163087
Gene Name
zinc finger protein 383
Gene Aliases
HSD17
Chromosome Location
Chr. 19: 37217243 - 37248740 on Build GRCh38
UniGene ID
Hs.590975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152604.1 NP_689817.1 5 672
XM_005258585.2 XP_005258642.1 6 930
XM_005258587.2 XP_005258644.1 5 807
XM_005258588.2 XP_005258645.1 6 569
XM_011526586.2 XP_011524888.1 7 1675
XM_011526587.2 XP_011524889.1 6 1642
XM_011526588.2 XP_011524890.1 5 1824
XM_011526589.2 XP_011524891.1 5 505
XM_011526590.2 XP_011524892.1 4 470
XM_017026422.1 XP_016881911.1 7 1081
XM_017026423.1 XP_016881912.1 7 1675
XM_017026424.1 XP_016881913.1 5 721
GenBank mRNA AK091760.1 - 5 1233
AK093182.1 - 5 672
AY260141.1 - 4 467
AY438646.1 - 5 672

View Full Product Details