Gene Transcripts

Gene Symbol
GTSCR1
Entrez Gene ID
220158
Gene Name
Gilles de la Tourette syndrome chromosome region, candidate 1 (non-protein coding)
Gene Aliases
Chromosome Location
Chr. 18: 70630519 - 70650857 on Build GRCh38
UniGene ID
Hs.448217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278515.1 NP_001265444.1 2 402
GenBank mRNA AY262164.1 - 2 402

View Full Product Details