Gene Transcripts
- Gene Symbol
- MBD3L4
- Entrez Gene ID
- 653656
- Gene Name
- methyl-CpG binding domain protein 3 like 4
- Gene Aliases
- Chromosome Location
- Chr. 19: 7037744 - 7040173 on Build GRCh38
- UniGene ID
- Hs.723810
- Species
- Homo sapiens
- Gene Symbol
- MBD3L2
- Entrez Gene ID
- 125997
- Gene Name
- methyl-CpG binding domain protein 3 like 2
- Gene Aliases
- Chromosome Location
- Chr. 19: 7049340 - 7051735 on Build GRCh38
- UniGene ID
- Hs.567667
- Species
- Homo sapiens
- Gene Symbol
- MBD3L3
- Entrez Gene ID
- 653657
- Gene Name
- methyl-CpG binding domain protein 3 like 3
- Gene Aliases
- Chromosome Location
- Chr. 19: 7056205 - 7058634 on Build GRCh38
- UniGene ID
- Hs.448960
- Species
- Homo sapiens
- Gene Symbol
- MBD3L5
- Entrez Gene ID
- 284428
- Gene Name
- methyl-CpG binding domain protein 3 like 5
- Gene Aliases
- Chromosome Location
- Chr. 19: 7030583 - 7033011 on Build GRCh38
- UniGene ID
- Hs.717312
- Species
- Homo sapiens
View Full Product Details