Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144606.5 | NP_653207.1 | 7 | 1191 | |
NM_144997.5 | NP_659434.2 | 7 | 1191 | ||
XM_011523714.2 | XP_011522016.1 | 8 | 1242 | ||
XM_011523716.1 | XP_011522018.1 | 6 | 1620 | ||
XM_011523718.2 | XP_011522020.1 | 7 | 1126 | ||
XM_011523719.2 | XP_011522021.1 | 8 | 1242 | ||
XM_011523721.2 | XP_011522023.1 | 7 | 991 | ||
XM_017024304.1 | XP_016879793.1 | 9 | 1411 | ||
XM_017024305.1 | XP_016879794.1 | 9 | 1426 | ||
XM_017024306.1 | XP_016879795.1 | 8 | 1415 | ||
XM_017024307.1 | XP_016879796.1 | 8 | 1377 | ||
XM_017024308.1 | XP_016879797.1 | 9 | 1553 | ||
XM_017024309.1 | XP_016879798.1 | 6 | 966 | ||
GenBank mRNA | AF517523.1 | - | 7 | 1142 | |
AL831885.1 | - | 1 | 530 | ||
BC015687.2 | - | 7 | 1142 | 4684974 | |
KU178888.1 | - | 4 | 687 | ||
KU178889.1 | - | 3 | 465 |