Gene Transcripts

Gene Symbol
SLC25A38
Entrez Gene ID
54977
Gene Name
solute carrier family 25 member 38
Gene Aliases
SIDBA2
Chromosome Location
Chr. 3: 39383324 - 39397328 on Build GRCh38
UniGene ID
Hs.369615
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017875.2 NP_060345.2 6 1117
XM_006713214.1 XP_006713277.1 7 1080
XM_011533869.2 XP_011532171.1 7 1980
GenBank mRNA AK026356.1 - 2 110
AK225388.1 - 6 1069
BC013194.1 - 6 778 3918744
CR457242.1 - 6 716

View Full Product Details