Gene Transcripts

Gene Symbol
GYPE
Entrez Gene ID
2996
Gene Name
glycophorin E (MNS blood group)
Gene Aliases
GPE, MNS, MiIX
Chromosome Location
Chr. 4: 143870866 - 143912291 on Build GRCh38
UniGene ID
Hs.654368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017008139.1 XP_016863628.1 1 493
XM_017008140.1 XP_016863629.1 1 493

View Full Product Details