Gene Transcripts

Gene Symbol
MUC3A
Entrez Gene ID
4584
Gene Name
mucin 3A, cell surface associated
Gene Aliases
MUC-3A, MUC3
Chromosome Location
Chr. 7: 100949054 - 100968347 on Build GRCh38
UniGene ID
Hs.703577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005960.1 NP_005951.1 2 2278
XM_011516246.2 XP_011514548.1 2 2637
XM_011516247.2 XP_011514549.1 2 2638
XM_011516249.2 XP_011514551.1 2 2642
XM_011516250.2 XP_011514552.1 2 2637
XM_017012231.1 XP_016867720.1 2 2644

View Full Product Details