Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172117.1 | NP_001165588.1 | 13 | 1728 | |
NM_010407.4 | NP_034537.2 | 13 | 1728 | ||
GenBank mRNA | AK149736.1 | - | 13 | 1694 | |
AK150290.1 | - | 13 | 1702 | ||
AK150709.1 | - | 13 | 1711 | ||
AK152506.1 | - | 13 | 1701 | ||
AK155975.1 | - | 13 | 1641 | ||
AK159234.1 | - | 13 | 1661 | ||
AK165315.1 | - | 13 | 1693 | ||
BC010478.1 | - | 13 | 1724 | 3978518 | |
BC013260.1 | - | 1 | 107 | ||
J03023.1 | - | 13 | 1617 | ||
Y00487.1 | - | 13 | 1623 |