Gene Transcripts
- Gene Symbol
- Klra10
- Entrez Gene ID
- 16628
- Gene Name
- killer cell lectin-like receptor subfamily A, member 10
- Gene Aliases
- Ly49J, Ly49i2
- Chromosome Location
- Chr. 6: 130266495 - 130284998 on Build GRCm38
- UniGene ID
- Mm.445916
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008459.2 |
NP_032485.2
|
1, 2 |
123 |
|
GenBank mRNA |
AF110492.1 |
- |
1, 2 |
123 |
|
View Full Product Details