Gene Transcripts

Gene Symbol
Lalba
Entrez Gene ID
16770
Gene Name
lactalbumin, alpha
Gene Aliases
AW208827
Chromosome Location
Chr. 15: 98480400 - 98482934 on Build GRCm38
UniGene ID
Mm.448567
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010679.1 NP_034809.1 2 232
GenBank mRNA BC006922.1 - 2 244
BC069916.1 - 2 232 3670536
M80909.1 - 2 232

View Full Product Details