Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297606.1 | NP_001284535.1 | 3 | 500 | |
NM_001297607.1 | NP_001284536.1 | 3 | 500 | ||
NM_009010.5 | NP_033036.2 | 3 | 500 | ||
GenBank mRNA | AK038300.1 | - | 3 | 477 | |
AK045224.1 | - | 3 | 490 | ||
AK161875.1 | - | 3 | 477 | ||
AK165467.1 | - | 3 | 501 | ||
BC003846.1 | - | 3 | 488 | ||
BC138684.1 | - | 3 | 460 | 8861705 | |
BC145372.1 | - | 3 | 460 | 9053930 | |
X92410.1 | - | 3 | 400 |