Gene Transcripts

Gene Symbol
Sat1
Entrez Gene ID
20229
Gene Name
spermidine/spermine N1-acetyl transferase 1
Gene Aliases
AA617398, SSAT, Sat
Chromosome Location
Chr. X: 155213126 - 155216449 on Build GRCm38
UniGene ID
Mm.2734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291865.1 NP_001278794.1 7 802
NM_009121.4 NP_033147.1 6 692
GenBank mRNA AK002531.1 - 6 593
AK131657.1 - 3 979
AK154445.1 - 6 588
BC058696.1 - 6 652 6820169
BC108334.1 - 6 1003
L10244.1 - 6 500

View Full Product Details