Gene Transcripts

Gene Symbol
Grk1
Entrez Gene ID
24013
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
Gprk1, Rhok, Rk
Chromosome Location
Chr. 8: 13405081 - 13421951 on Build GRCm38
UniGene ID
Mm.257501
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011881.3 NP_036011.3 1 607
GenBank mRNA BC075722.1 - 1 582
BC096611.1 - 1 607 5357325

View Full Product Details