Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020028.3 | NP_064412.2 | 3 | 1159 | |
GenBank mRNA | AK051092.1 | - | 3 | 1109 | |
AK144861.1 | - | 3 | 1160 | ||
BC060131.1 | - | 3 | 1295 | 6409729 | |
BC064676.1 | - | 3 | 1295 | 6409729 |