Gene Transcripts
- Gene Symbol
- LGALS4
- Entrez Gene ID
- 3960
- Gene Name
- galectin 4
- Gene Aliases
- GAL4, L36LBP
- Chromosome Location
- Chr. 19: 38801671 - 38813544 on Build GRCh38
- UniGene ID
- Hs.5302
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006149.3 |
NP_006140.1
|
9, 10 |
1033 |
|
XM_011526973.1 |
XP_011525275.1
|
8, 9 |
1438 |
|
XM_011526974.1 |
XP_011525276.1
|
8, 9 |
1272 |
|
GenBank mRNA |
AB006781.1 |
- |
9, 10 |
875 |
|
AF014838.1 |
- |
9, 10 |
832 |
|
AK290269.1 |
- |
9, 10 |
878 |
|
BC003661.2 |
- |
9, 10 |
862 |
2967411 |
BC005146.2 |
- |
9, 10 |
862 |
2988384 |
BC034750.1 |
- |
9, 10 |
858 |
5183145 |
BT007032.1 |
- |
9, 10 |
819 |
|
CR536544.1 |
- |
9, 10 |
819 |
|
U82953.1 |
- |
9, 10 |
842 |
|
View Full Product Details