Gene Transcripts
- Gene Symbol
- Sirt2
- Entrez Gene ID
- 64383
- Gene Name
- sirtuin 2
- Gene Aliases
- 5730427M03Rik, SIR2L2, Sir2l
- Chromosome Location
- Chr. 7: 28766752 - 28788665 on Build GRCm38
- UniGene ID
- Mm.272443
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122765.1 |
NP_001116237.1
|
5, 6 |
435 |
|
NM_001122766.1 |
NP_001116238.1
|
3, 4 |
272 |
|
NM_022432.4 |
NP_071877.3
|
6, 7 |
482 |
|
GenBank mRNA |
AB221570.1 |
- |
6, 7 |
387 |
|
AF299337.1 |
- |
6, 7 |
467 |
|
AK014042.1 |
- |
5, 6 |
406 |
|
AK144431.1 |
- |
5, 6 |
378 |
|
AK146412.1 |
- |
5, 6 |
415 |
|
BB868649.1 |
- |
3, 4 |
237 |
|
BC021439.1 |
- |
6, 7 |
486 |
5039278 |
KF032392.1 |
- |
6, 7 |
424 |
|
View Full Product Details