Gene Transcripts

Gene Symbol
LOR
Entrez Gene ID
4014
Gene Name
loricrin
Gene Aliases
Chromosome Location
Chr. 1: 153259703 - 153262124 on Build GRCh38
UniGene ID
Hs.251680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000427.2 NP_000418.2 2 1146
GenBank mRNA BC034690.1 - 1 1137
BC108290.1 - 2 1158 4754528
M61120.1 - 1 1137

View Full Product Details