Gene Transcripts
- Gene Symbol
- Fam103a1
- Entrez Gene ID
- 67148
- Gene Name
- family with sequence similarity 103, member A1
- Gene Aliases
- 2410047I02Rik, 2610204K14Rik, RAM
- Chromosome Location
- Chr. 7: 81762953 - 81769490 on Build GRCm38
- UniGene ID
- Mm.241472
- Species
- Mus musculus
- Gene Symbol
- Gm10767
- Entrez Gene ID
- 100038538
- Gene Name
- predicted gene 10767
- Gene Aliases
- ENSMUSG00000074826
- Chromosome Location
- Chr. 13: 66905415 - 66909166 on Build GRCm38
- UniGene ID
- Mm.405005
- Species
- Mus musculus
View Full Product Details