Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000243.2 | NP_000234.1 | 2 | 754 | |
XM_017023236.1 | XP_016878725.1 | 2 | 895 | ||
GenBank mRNA | AF018080.1 | - | 2 | 755 | |
AK298660.1 | - | 2 | 748 | ||
BC101511.1 | - | 2 | 739 | 8069017 | |
BC101537.1 | - | 2 | 739 | 8069043 | |
FJ785721.1 | - | 2 | 714 | ||
Y14441.1 | - | 2 | 743 |