Gene Transcripts

Gene Symbol
MTNR1B
Entrez Gene ID
4544
Gene Name
melatonin receptor 1B
Gene Aliases
FGQTL2, MEL-1B-R, MT2
Chromosome Location
Chr. 11: 92969623 - 92986241 on Build GRCh38
UniGene ID
Hs.569039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005959.3 NP_005950.1 1, 2 334
XM_011542839.2 XP_011541141.1 1, 2 231
GenBank mRNA AY521019.1 - 1, 2 231
BC069163.1 - 1, 2 334 7216982
U25341.1 - 1, 2 243

View Full Product Details