Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028074.4 | NP_082350.3 | 16, 17 | 2272 | |
GenBank mRNA | AK007805.1 | - | 3, 4 | 520 | |
AK049311.1 | - | 16, 17 | 2274 | ||
AK134182.1 | - | 15, 16 | 2078 | ||
AK169816.1 | - | 16, 17 | 2218 | ||
AK171730.1 | - | 16, 17 | 2141 | ||
BC043036.1 | - | 15, 16 | 2029 | 5698965 | |
BC049382.1 | - | 4, 5 | 684 |