Gene Transcripts

Gene Symbol
Nudt9
Entrez Gene ID
74167
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 9
Gene Aliases
1190002C07Rik, ADPR-PPase, AI462474
Chromosome Location
Chr. 5: 104046514 - 104065369 on Build GRCm38
UniGene ID
Mm.241484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004444.1 - 6 755
AK076500.1 - 6 742
AK151245.1 - 5 561
AK158058.1 - 6 817
AK219306.1 - 1 45

View Full Product Details